Tin(II)chlorid 30% in hydrochloric acid 20% Reag. Ph. Eur. 1085001

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Stannous chloride, Tin dichloride

87941.180EA 795 DKK
87941.180
Tin(II)chlorid 30% in hydrochloric acid 20% Reag. Ph. Eur. 1085001
Tin(II)chlorid

Bestillingsoplysninger: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: SnCl₂
MDL Number: MFCD00011241
CAS-nummer: 7772-99-8
EINECS: 231-868-0
UN nummer: 1789
ADR: 8,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR