Natriumcarbonat 2% w/v sodium hydroxide solution 0.1 mol/l Reag. Ph. Eur. 1079302

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Calcined soda, Carbonic acid disodium salt

87929.290EA 1194 DKK
87929.290
Natriumcarbonat 2% w/v sodium hydroxide solution 0.1 mol/l Reag. Ph. Eur. 1079302
Sodium carbonate

Bestillingsoplysninger: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: Na₂CO₃
Molekylvægt: 105.99 g/mol
Kogepunkt: 1600
Smeltepunkt: 854
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003494
CAS-nummer: 497-19-8
EINECS: 207-838-8
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR