1,4-Dioxan 0,1% aqueous solution Reag. Ph. Eur. 1032001

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: p-Dioxan

87820.180EA 876 DKK
87820.180
1,4-Dioxan 0,1% aqueous solution Reag. Ph. Eur. 1032001
1,4-Dioxan

Bestillingsoplysninger: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: C₄H₈O₂
Molekylvægt: 88.11 g/mol
Kogepunkt: 101.1
Smeltepunkt: 11.8
Flammepunkt: 12
MDL Number: MFCD00006571
CAS-nummer: 123-91-1
EINECS: 204-661-8
UN nummer: 1165
ADR: 3 II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR