Anisol ≥99%, GPR RECTAPUR®

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methyl phenyl ether, Methoxybenzene

87106.180EA 337 DKK
87106.180 87106.260 87106.290 87106.320 87106.460
Anisol ≥99%, GPR RECTAPUR®
Anisol
Formel: C₇H₈O
Flammepunkt: 52 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008354
CAS-nummer: 100-66-3
UN nummer: 2222
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Assay ≥ 99 %
Appearance Clear colourless liquid
Odour Characteristic
Density (20/4) 0,992 - 0,994


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR