Kobber 99,7, powder_fine, Analar NORMAPUR® ≤63 µm analysis_for

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

87059.230EA 0 DKK
87059.230 87059.290
Kobber 99,7, powder_fine, Analar NORMAPUR® ≤63 µm analysis_for
Kobber
Formel: Cu
Molekylvægt: 63.55 g/mol
Kogepunkt: 2595
Smeltepunkt: 1083
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010965
CAS-nummer: 7440-50-8
UN nummer: 3089
ADR: 4.1 III
Merck Index: 13,02545

Bestil nu

Specifikationer Testresultater

Assay Min. 99,7 %
Particle size (< 45 µm) Min. 90 %
Particle size (> 75 µm) Max. 1 %
Insolubility in nitric acid Max. 0,02 %
P (Phosphorus) Max. 10 ppm
Ag (Silver) Max. 20 ppm
As (Arsenic) Max. 1 ppm
Fe (Iron) Max. 50 ppm
Mn (Manganese) Max. 10 ppm
Pb (Lead) Max. 100 ppm
Sb (Antimony) Max. 10 ppm
Sn (Tin) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR