Urea 98.0-102% USP, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Urinstof

8642-30EA 36520 DKK
8642-30 8642-12 8642-88
Urea 98.0-102% USP, Macron Fine Chemicals™
Urea
Formel: NH₂CONH₂
Molekylvægt: 60.06 g/mol
Kogepunkt: 197 °C (1013 hPa)
Smeltepunkt: 132.7 °C
Massefylde: 1.34 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008022
CAS-nummer: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

GMP Manufactured Product
Meets U.S.P Requirements
Preserve in tight containers. Store at 25°C, excursions permitted 15-30°C.
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (NH₂CONH₂) 98.0 - 102.0 %
USP - Alcohol Insoluble Matter ≤0.04 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Residue on Ignition ≤0.1 %
USP - Organic impurities: Urea related compound A ≤0.1 %
USP - Organic impurities: Any individual unspecified impurity ≤0.1 %
USP - Organic impurities: Total impurities ≤2.0 %
Appearance (colorless to white powder, granules, or small pellets)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR