Kaliumdichromat solution_SC 0.02 mol/l (0.12 N), AVS TITRINORM COD_determination

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

86181.290EA 324 DKK
86181.290
Kaliumdichromat solution_SC 0.02 mol/l (0.12 N), AVS TITRINORM COD_determination
Potassium dichromate

Certificeringer: The production takes place according to DIN 8409:1980, DEV H41 DIN 38409:1981, DEV H43.

Formel: K₂Cr₂O₇
Molekylvægt: 294.18 g/mol
Kogepunkt: 500
Smeltepunkt: 398
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011367
CAS-nummer: 7778-50-9
UN nummer: 3287
ADR: 6.1 III
Merck Index: 12,07790

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR