Hav sand, GPR RECTAPUR®

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

86154.295EA 592 DKK
86154.295 86154.364 86154.460
Hav sand, GPR RECTAPUR®
Hav sand
Formel: SiO₂
Molekylvægt: 60.08 g/mol
Kogepunkt: 2230
Smeltepunkt: 1710
CAS-nummer: 14808-60-7

Bestil nu

Specifikationer Testresultater

Substances soluble in HCl max 0.5%
Chloride (Cl) max. 0.01%
Loss on ignition (900 °C) max. 0.1%
Particle size (0,1 - 0,315 mm) min.90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR