Ethanol absolute ≥99.8%, PESTINORM® for pesticide residue analysis

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ethyl alcohol

85933.290EA 562 DKK
85933.290 85933.320
Ethanol absolute ≥99.8%, PESTINORM® for pesticide residue analysis
Ethanol
Formel: C₂H₅OH
Molekylvægt: 46.07 g/mol
Kogepunkt: 78.3 °C (1013 hPa)
Smeltepunkt: –117 °C
Flammepunkt: 12
MDL Number: MFCD00003568
CAS-nummer: 64-17-5
EINECS: 200-578-6
UN nummer: 1170
ADR: 3 II
Merck Index: 12,03806

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless liquid
Assay (GC, on anhydrous basis) min. 99.8 %
Identity (IR) Comply
Residue after evaporation max. 0.0003 % w/w
Water (KF) max. 0.05 % w/w
GC/ECD any Pesticide (as Lindane) max. 5 pg/ml

Specifikationer

Environmentally Preferable Sikrere sundhed for mennesker og miljø
Miljøcertificering EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR