Trifluoreddikesyre ≥99.8% (by anhydrous basis), SPECTRONORM® for spectroscopy

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: TFA

Packaged under nitrogen.

85930.150EA 366 DKK
85930.150 85930.180
Trifluoreddikesyre ≥99.8% (by anhydrous basis), SPECTRONORM® for spectroscopy
Trifluoreddikesyre

SPECTRONORM solvents have extensive specifications to meet the highly demanding requirements of UV applications.

Formel: C₂HF₃O₂
Molekylvægt: 114.02 g/mol
Kogepunkt: 71.8
Smeltepunkt: –15 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004169
CAS-nummer: 76-05-1
UN nummer: 2699
ADR: 8 I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay (on anhydrous substance) min 99.8%
Water max 0.10%
Colour number (APHA) max 10
Evaproration residue max 0.005%
Transmittance (at 265 nm) min 10%
Transmittance (at 305 nm) min 50%
Transmittance (at 320 nm) min 80%
Transmittance (at 325 nm) min 90%
manufactured and filled under nitrogen Yes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR