Copper test strips, Dosatest
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
85696.601EA 216 DKK
85696.601
Copper test strips, Dosatest
Miljø test kit
Semi-quantitative test strips for the determination of copper.
  • Ideal for rapid and easy analysis of copper directly on-site
  • No maintenance or calibration is required
Colour change: White to red-violet.
Leveringsinformation: Delivered as 100 test strips in a tube with instruction leaflet.
Advarsel: Strongly acid solutions (pH < 2) have to be buffered to a pH value of 2 to 6 using solid sodium acetate.