Myresyre ≥90%, AnalaR NORMAPUR® for analysis of viscosity following DIN EN ISO 307

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Aminic Acid, Methanoic acid

85494.320EA 862 DKK
85494.320
Myresyre ≥90%, AnalaR NORMAPUR® for analysis of viscosity following DIN EN ISO 307
Myresyre
Formel: HCO₂H
Molekylvægt: 46.03 g/mol
Massefylde: 1.208 g/cm³ (20 °C)
Flammepunkt: 48
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003297
CAS-nummer: 64-18-6
EINECS: 0
UN nummer: 1779
ADR: 8 (3) II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay (at filling) 90.00 - 90.15 %
Density (20/4) (at filling) 1.2044 - 1.2047
Acetic acid ≤ 0.2 %
Conforms to ISO 307 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR