Acetone 99,9, PESTINORM® Supra Trace Trace_analysis_organic

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Propanon

Filter 0,2 um_Inert_Gas_filled

85384.320EA 921 DKK
85384.320
Acetone 99,9, PESTINORM® Supra Trace Trace_analysis_organic
Acetone
Formel: CH₃COCH₃
Molekylvægt: 58.08 g/mol
Kogepunkt: 56.2 °C (1013 hPa)
Smeltepunkt: –95.4 °C
Massefylde: 0.792 g/cm³ (20 °C)
Flammepunkt: <–20 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008765
CAS-nummer: 67-64-1
EINECS: 200-662-2
UN nummer: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Clear colourless liquid
Identity (IR) Conforms
Assay (calculated on anhydrous) Min. 99.9 %
Residue after evaporation Max. 0.0003 %(m)
Water Max. 0.3 %(m)
Colour Max. 10 APHA
GC/ECD dioxins, furans and PCB's Max. 5 ng/l
GC/ECD any pesticide (as Lindane) Max. 5 ng/l
GC/NPD any pesticide (as Parathion) Max. 10 ng/l
GC/ECD 1,2,4-TCB to deca-PCB(as Lindane) Max. 5 pg/ml
GC/ECD DCM to 1,2,4-TCB (as TCM) Max. 2.0 ng/ml
GC/FID C10 to C40 (as n-Decane) Max. 2.0 ng/ml
GC/MSD C10 to C40 (as Decane; 30-600amu) Max. 2.0 ng/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR