Sølvnitrat 0.1 N in aqueous solution USP Test Solution (TS)

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Lunar caustic, Silver mononitrate, Nitric acid silver salt

85370.180EA 1360 DKK
85370.180 85370.260
Sølvnitrat 0.1 N in aqueous solution USP Test Solution (TS)
Sølvnitrat

Ready to use solution.


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Formel: AgNO₃
Molekylvægt: 169.87 g/mol
Kogepunkt: 444
Smeltepunkt: 210
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003414
CAS-nummer: 7761-88-8
EINECS: 0
UN nummer: 3264
ADR: 8 III
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR