Methanol med 0,1% (v/v) Trifluoreddikesyre, HiPerSolv CHROMANORM® for LC-MS

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig


Packaged under nitrogen.

84870.290EA 682.5 DKK
84870.290 84870.320
Methanol med 0,1% (v/v) Trifluoreddikesyre, HiPerSolv CHROMANORM® for LC-MS
Methanol med 0,1% (v/v) Trifluoreddikesyre
Opbevaringstemperatur: Stue temperatur
UN nummer: 1230
ADR: 3 (6.1) II

Bestil nu

Specifikationer Testresultater

Nominal concentration (V/V) 0.095 - 0.105 %
Evaporation residue Max. 0.0001 %
Water Max. 0.02 %
Ca (Calcium) Max. 0.2 ppm
K (Potassium) Max. 0.2 ppm
Mg (Magnesium) Max. 0.2 ppm
Na (Sodium) Max. 0.5 ppm
Transmittance (225 nm) Min. 20 %
Transmittance (240 nm) Min. 50 %
Transmittance (250 nm) Min. 80 %
Transmittance (260 nm) Min. 95 %
Transmittance (270 nm) Min. 99 %
Suitable for LC-MS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR