2-Propanol, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Isopropylalkohol

8468.2500EA 1330 DKK
8468.2500
2-Propanol, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
2-Propanol
Formel: (CH₃)₂CHOH
MDL Number: MFCD00011674
CAS-nummer: 67-63-0
UN nummer: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Assay (by GC) min. 99.8 %
Acidity max. 0.0005 meq/g
Evaporation Residue max. 5 ppm
GC sensitive impurities (lindane GC-ECD) max. 5 ng/l
Water (KF) max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR