Natriumhydroxid concentrate for dilution to 0.5 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Caustic soda

Dose for preparation of 1 litre of solution.

84597.600EA 207 DKK
84597.600
Natriumhydroxid concentrate for dilution to 0.5 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Sodium hydroxide
Formel: NaOH
Molekylvægt: 40 g/mol
Kogepunkt: >105 °C (1013 hPa)
Smeltepunkt: 323
Massefylde: 1.327 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
UN nummer: 1824
ADR: 8 II
Merck Index: 14,08627

Bestil nu

Specifikationer Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.4998 0.5002 mol/l

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR