Dichlormethan 99,9 Stabilised, PESTINORM® GC_capillary
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Methylenchlorid
Stabilization: 20 ppm 2-Methyl-2-buten
Filter 0,2 um_Nitrogen_filled
83961.320EA 1250 DKK
83961.320
Dichlormethan 99,9 Stabilised, PESTINORM® GC_capillary
Dichloromethane
Formel: CH₂Cl₂
Molekylvægt: 84.93 g/mol
Kogepunkt: 39.8 °C (1013 hPa)
Smeltepunkt: –95 °C
Massefylde: 1.322 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000881
CAS-nummer: 75-09-2
EINECS: 200-838-9
UN nummer: 1593
ADR: 6.1 III
REACH: 01-2119480404-41
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Assay (on anhydrous substance) Min. 99.9 %
Acidity Max. 0.0005 meq/g
Evaporation residue (100°C) Max. 0.0005 %
Water Max. 0.05 %
Organic residue (as Octanol) (GC/FID) Max. 10 ng/l
Halogenated residue (as Lindane)(GC/ECD) Max. 5 ng/l