Ethanol absolute ≥99.5%, TechniSolv®, pure

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethyl alcohol

83813.360_TEA 381 DKK
83813.360_T 83813.410_T 83813.440_T 83813.360 83813.410 83813.440 83813.550 83813.930
Ethanol absolute ≥99.5%, TechniSolv®, pure
Ethanol

Advarsel: WARNING : This alcohol is regulated. Please contact VWR.

Formel: H₃CCH₂OH
Molekylvægt: 46.07 g/mol
Kogepunkt: 78.3 °C (1013 hPa)
Smeltepunkt: –117 °C
Massefylde: 0.7895 g/cm³ (20 °C)
Flammepunkt: 12 °C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003568
CAS-nummer: 64-17-5
EINECS: 200-578-6
UN nummer: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Bestil nu

Specifikationer Testresultater

Assay (V/V) Min. 99.5 %
Density (20/20) 0.788 - 0.794
Water Max. 0.5 %

Specifikationer

Environmentally Preferable Sikrere sundhed for mennesker og miljø Sikrere sundhed for mennesker og miljø
Miljøcertificering EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR