Ethanol 90% 90% (v/v), TechniSolv®, pure

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethyl alcohol

83811.410EA 1070 DKK
83811.410 83811.360
Ethanol 90% 90% (v/v), TechniSolv®, pure
Ethanol

Advarsel: WARNING : this alcohol is regulated. Call us.

Formel: C₂H₅OH
Molekylvægt: 46.07 g/mol
Kogepunkt: 78.3
Smeltepunkt: -117
Flammepunkt: 12
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003568
CAS-nummer: 64-17-5
EINECS: 200-578-6
UN nummer: 1170
ADR: 3 II
Merck Index: 12,03806

Bestil nu

Specifikationer Testresultater

Assay Min. 89.5 %
Density (20/20) 0.827 - 0.833 kg/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR