Kviksølv(II)sulfat ≥98.0%, AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Mercury sulphate, Mercuric sulphate

83565.180EA 2070 DKK
83565.180 83565.230
Kviksølv(II)sulfat ≥98.0%, AnalaR NORMAPUR® analytical reagent
Kviksølv(II)sulfat
Formel: HgSO₄
Molekylvægt: 296.65 g/mol
Smeltepunkt: 850
Massefylde: 6.47 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011047
CAS-nummer: 7783-35-9
EINECS: 231-992-5
UN nummer: 1645
ADR: 6.1 II
Merck Index: 13,05914

Bestil nu

Specifikationer Testresultater

Assay Min. 98.0 %
Chemical oxygen demand (COD) Passes test
Residue after reduction Max. 0.1 %
Cl (Chloride) Max. 30 ppm
NO3 (Nitrate) Max. 50 ppm
Fe (Iron) Max. 50 ppm
Hg (I) (Mercury) Max. 0.15 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR