Magnesiumoxid heavy 98,0-100,5 Ph. Eur.

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Magnesia usta

83540.290EA 611 DKK
83540.290
Magnesiumoxid heavy 98,0-100,5 Ph. Eur.
Magnesium oxide
Formel: MgO
Molekylvægt: 40.3 g/mol
Kogepunkt: 3600
Smeltepunkt: 2800 °C
Massefylde: 3.58 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011109
CAS-nummer: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Bestil nu

Specifikationer Testresultater

Assay (calculated on ignited substance) 98.0 - 100.5 %
Appearance Conforms (see CoA/CoS)
Untapped bulk density ≥ 0.25 g/ml
Identification (Mg) Passes test
Appearance of solution Passes test
Soluble substances ≤ 2.0 %
Substances insoluble in CH3COOH ≤ 0.1 %
Cl (Chloride) ≤ 0.1 %
SO4 (Sulphate) ≤ 1.0 %
Ca (Calcium) ≤ 1.5 %
Fe (Iron) ≤ 0.07 %
Heavy metals (as Pb) ≤ 30 ppm
Loss on ignition (900°C) ≤ 8.0 %
Residual solvents Passes test
As (Arsenic) ≤ 4 ppm
Cd (Cadmium) ≤ 1 ppm
Co (Cobalt) ≤ 1 ppm
Hg (Mercury) ≤ 2 ppm
Ni (Nickel) ≤ 3 ppm
Pb (Lead) ≤ 3 ppm
V (Vanadium) ≤ 1 ppm
Conforms to Ph.Eur. Passes test

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR