Diethylether stabilised, BAKER, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

8106.1000EA 425 DKK
8106.1000 8106.5000 8106.9025 8106.9200
Diethylether stabilised, BAKER, J.T.Baker®
Diethylether
Formel: (CH₃CH₂)₂O
Molekylvægt: 74.12 g/mol
Kogepunkt: 35
Smeltepunkt: –123 °C
Massefylde: 0.7135 g/cm³ (20 °C)
Flammepunkt: –40 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011646
CAS-nummer: 60-29-7
EINECS: 200-467-2
UN nummer: 1155
ADR: 3 I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Meets Reagent Specifications for testing Ph. Eur. monographs.
Acidity passes test
Aldehydes passes test
Distillation Range 34.0 - 35.0 °C
Non-volatile Matter max. 20 mg/l
Peroxide passes test
Relative Density 0.714 - 0.716
Substances with a Foreign Odour passes test
Water (H2O) max. 2 g/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR