Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ethyl alcohol

8025.1000EA 306 DKK
8025.1000 8025.0250 8025.9025 8025.5000PE 8025.2500PE 8025.2500 8025.1000PE 8025.9200
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Ethanol
Formel: C₂H₅OH
Molekylvægt: 46.07 g/mol
Kogepunkt: 78.3
Smeltepunkt: –117 °C
Massefylde: 0.7895 g/cm³ (20 °C)
Flammepunkt: 12
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003568
CAS-nummer: 64-17-5
EINECS: 200-578-6
UN nummer: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Bestil nu

Specifikationer Testresultater

Exceeds ACS Specifications
Assay min. 99.9 % (v/v)
Acetone, isopropylalcohol passes test
Color (APHA) max. 10
Methanol (CH3OH) max. 0.1 %
Residue after Evaporation max. 0.001 %
Solubility in Water passes test
Substances Darkened by H2SO4 passes test
Substances Reducing KMnO4 passes test
Titrable Acid (meq/g) max. 0.0005
Titrable Base (meq/g) max. 0.0002
Water (H2O) max. 0.2 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.2
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR