Thiourea, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Thiourinstof

M-Clarity™ quality level = MQ200

8.45501.9025 8.45501.9010 8.45501.9011 8.45501.9013
8.45501.9025EA 0 DKK
8.45501.9025 8.45501.9010 8.45501.9011 8.45501.9013
Thiourea, Sigma-Aldrich®
Thiourea
Formel: NH₂CSNH₂
Smeltepunkt: 174…177 °C
Massefylde: 1,435 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008067
CAS-nummer: 62-56-6
EINECS: 200-543-5
UN nummer: 3077
ADR: 9,III
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Hill Formula CH₄N₂S
Chemical formula H₂NCSNH₂
Molar Mass 76.12 g/mol
Density 1.405 g/cm³ (20 °C)
Melting Point 169 - 173 °C
pH value 6 - 8 (50 g/l, H₂O, 20 °C)
Vapor pressure (20 °C)
Bulk density 640 kg/m³
Solubility 137 g/l (20 °C)
LD 50 oral LD50 Rat 1750 mg/kg
LD 50 dermal LD50 Rabbit 2800 mg/kg
R Phrase R 22-40-51/53-63
S Phrase S 36/37-61


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR