Titan(IV)ethanolat for synthesis

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Titanium (IV) ethoxide, Titanium tetraethanolate, Tetraethyl orthotitanate

M-Clarity™ quality level = MQ200

8.21083.0250 8.21083.1000
8.21083.0250EA 662 DKK
8.21083.0250 8.21083.1000
Titan(IV)ethanolat for synthesis
Titan(IV)ethanolat
Formel: C₈H₂₀O₄Ti
Kogepunkt: 150 °C (1013 hPa)
Massefylde: 1,107 g/cm³ (20 °C)
Flammepunkt: 33 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00009071
CAS-nummer: 3087-36-3
EINECS: 221-410-8
UN nummer: 1993
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) Hydrolysis
Melting Point -40 °C
Molar Mass 228.15 g/mol
Boiling Point 150 - 155 °C (16 hPa)
Flash Point 33 °C
Refractive Index 1.5043 (20 °C, 589 nm)
Density 1.08 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR