Kaliumtetrahydroborat for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Potassium tetrahydridoborate, Potassium borohydride

M-Clarity™ quality level = MQ200

8.20747.0025 8.20747.0100 8.20747.1000
8.20747.0025EA 452 DKK
8.20747.0025 8.20747.0100 8.20747.1000
Kaliumtetrahydroborat for synthesis, Sigma-Aldrich®
Potassium tetrahydroborate
Formel: H₄BK
Smeltepunkt: 500 °C
Massefylde: 1,17 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011396
CAS-nummer: 13762-51-1
EINECS: 237-360-5
UN nummer: 1870
ADR: 4.3,I
Merck Index: 12,07777

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) (rigorous decomposition)
Melting Point 500 °C
Molar Mass 53.94 g/mol
Bulk Density 400 kg/m3
Density 1.11 g/cm3
Hazard Class 4.3
Hazard Class Description Dangerous When Wet Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR