Palladium(II)acetat zS, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Palladium diacetate, Palladium (II) acetate

MQ200

8.18056.0001 8.18056.0005 8.18056.8500
8.18056.0001EA 1900 DKK
8.18056.0001 8.18056.0005 8.18056.8500
Palladium(II)acetat zS, Sigma-Aldrich®
Palladium(II)acetat
Formel: C₄H₆O₄Pd
Molekylvægt: 224.51 g/mol
Smeltepunkt: 205
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012453
CAS-nummer: 3375-31-3
EINECS: 222-164-4

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) insoluble
Melting Point 205 °C
Molar Mass 224.5 g/mol
pH 2 - 3 (H2O, 20 °C) (aqueous suspension)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR