Natriumchlorit 25% in aqueous solution for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig


M-Clarity™ quality level = MQ200

8.14815.0250 8.14815.1000
8.14815.0250EA 333 DKK
8.14815.0250 8.14815.1000
Natriumchlorit 25% in aqueous solution for synthesis, Sigma-Aldrich®
Sodium chlorite
Formel: ClNaO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003478
CAS-nummer: 7758-19-2
EINECS: 231-836-6
UN nummer: 1908
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Boiling Point 100 °C
Density 1.21 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR