Natriumhydrogensulfit 39% in aqueous solution for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Sodium hydrogen sulphite

M-Clarity™ quality level = MQ200

8.06356.1000 8.06356.2500 8.06356.9025
8.06356.1000EA 326 DKK
8.06356.1000 8.06356.2500 8.06356.9025
Natriumhydrogensulfit 39% in aqueous solution for synthesis, Sigma-Aldrich®
Sodium bisulphite
Formel: HNaO₃S
MDL Number: MFCD00167602
CAS-nummer: 7631-90-5
EINECS: 231-548-0
UN nummer: 2693
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Solubility (20 °C) soluble
Melting Point -44 °C
Boiling Point 146 °C
Vapor Pressure 40 hPa (20 °C)
Density 1.34 g/cm3 (20 °C)
pH 4.5 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR