4-(Dimethylamino)benzaldehyd for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde

M-Clarity™ quality level = MQ200

8.03057.0100 8.03057.0250 8.03057.1000 8.03057.0025
8.03057.0100EA 432 DKK
8.03057.0100 8.03057.0250 8.03057.1000 8.03057.0025
4-(Dimethylamino)benzaldehyd for synthesis, Sigma-Aldrich®
4-(Dimethylamino)benzaldehyde
Formel: (CH₃)₂NC₆H₄CHO
Kogepunkt: 176…177 °C (1013 hPa)
Smeltepunkt: 72 °C
Massefylde: 1,0254 g/cm³ (99.9 °C)
Flammepunkt: >100 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003381
CAS-nummer: 100-10-7
EINECS: 202-819-0
UN nummer: 3077
ADR: 9,III
Merck Index: 13,03257
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Solubility 0.3 g/l (20 °C)
Melting Point 72 - 75 °C
Molar Mass 149.19 g/mol
Bulk Density 350 kg/m3
Boiling Point 176 - 177 °C (23 hPa)
Flash Point 100 °C
Ignition Point 445 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR