2-Amino-2-methylpropanol for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-2-methyl-1-propanol, Isobutanolamine, AMP

M-Clarity™ quality level = MQ200

8.01465.0250 8.01465.1000 8.01465.2500 8.01465.9026
8.01465.0250EA 346 DKK
8.01465.0250 8.01465.1000 8.01465.2500 8.01465.9026
2-Amino-2-methylpropanol for synthesis, Sigma-Aldrich®
2-Amino-2-methylpropanol
Formel: (CH₃)₂C(NH₂)CH₂OH
Kogepunkt: 165 °C (1013 hPa)
Smeltepunkt: 18…26 °C
Massefylde: 0,667 g/cm³ (20 °C)
Flammepunkt: 67 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008051
CAS-nummer: 124-68-5
EINECS: 204-709-8
Merck Index: 13,00448

Bestil nu

Specifikationer Testresultater

Melting Point 30 - 31 °C
Molar Mass 89.14 g/mol
Boiling Point 165 °C (1013 hPa) (anhydrous)
Vapor Pressure 0.45 hPa (20 °C)
Flash Point 77.8 °C
Refractive Index 1.448 (20 °C, 589 nm)
Density 0.95 g/cm3 (25 °C)
pH 11.3 (10 g/l, H2O)
Ignition Point 438 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR