Dimethylphthalat for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Phthalic acid dimethyl ester

M-Clarity™ quality level = MQ200

8.00918.0100 8.00918.1000 8.00918.2500
8.00918.0100EA 243 DKK
8.00918.0100 8.00918.1000 8.00918.2500
Dimethylphthalat for synthesis, Sigma-Aldrich®
Dimethylphthalat
Formel: C₁₀H₁₀O₄
Molekylvægt: 194.19 g/mol
Kogepunkt: 283.7
Smeltepunkt: 6
Massefylde: 1.192 g/cm³ (20 °C)
Flammepunkt: 146
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008425
CAS-nummer: 131-11-3
EINECS: 205-011-6
Merck Index: 13,03281

Bestil nu

Specifikationer Testresultater

Solubility  - 0.1 g/l (20 °C)
Melting Point 6 °C
Molar Mass 194.18 g/mol
Boiling Point 283 °C (1013 hPa)
Vapor Pressure 0.008 hPa (20 °C)
Flash Point 150 °C
Refractive Index 1.5137 (25 °C, 589 nm)
Density 1.19 g/cm3 (20 °C)
pH 7 (H2O, 20 °C)
Ignition Point 460 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR