Chloreddikesyre for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Monochloroacetic acid

M-Clarity™ quality level = MQ200

8.00412.0100 8.00412.1000 8.00412.2500 8.00412.9050 8.00412.9025
8.00412.0100EA 275 DKK
8.00412.0100 8.00412.1000 8.00412.2500 8.00412.9050 8.00412.9025
Chloreddikesyre for synthesis, Sigma-Aldrich®
Chloreddikesyre
Formel: ClCH₂COOH
Kogepunkt: 188 °C (1013 hPa)
Smeltepunkt: 56…57 °C
Massefylde: 1,4043 g/cm³ (20 °C)
Flammepunkt: 126 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002683
CAS-nummer: 79-11-8
EINECS: 201-178-4
UN nummer: 1751
ADR: 6.1 (8),II
Merck Index: 13,02129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2516e0c26c6c26f4f6f3656bc5b4256e7cf08207&vid=9c9b7ad094469d9089eb2f0ce88275d1423c2b46&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Solubility 4210 g/l (20 °C)
Melting Point 62 °C
Molar Mass 94.49 g/mol
Bulk Density 800 kg/m3
Boiling Point 189 °C (1013 hPa)
Vapor Pressure 1 hPa (20 °C)
Flash Point 126 °C
Explosion Limit 8.0 %(V)
Density 1.37 g/m3 (65 °C)
pH  - 1 (800 g/l, H2O, 20 °C)
Ignition Point 460 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR