Fumarsyre for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: trans-1,2-Ethylentricarboxylic acid

M-Clarity™ quality level = MQ200

8.00269.0100 8.00269.1000 8.00269.2500 8.00269.9050
8.00269.0100EA 176 DKK
8.00269.0100 8.00269.1000 8.00269.2500 8.00269.9050
Fumarsyre for synthesis, Sigma-Aldrich®
Fumarsyre
Formel: C₄H₄O₄
Molekylvægt: 116.07 g/mol
Kogepunkt: 356
Smeltepunkt: 287
Massefylde: 1.625 g/cm³ (20 °C)
Flammepunkt: 273
MDL Number: MFCD00002700
CAS-nummer: 110-17-8
EINECS: 203-743-0
Merck Index: 13,04308

Bestil nu

Specifikationer Testresultater

Solubility 4.9 g/l (20 °C)
Melting Point 287 °C
Molar Mass 116.07 g/mol
Boiling Point 290 °C (1013 hPa) (sublimed)
Vapor Pressure  - 0.001 hPa (20 °C)
Flash Point 230 °C
Density 1.64 g/cm3 (20 °C)
pH 2.1 (4.9 g/l, H2O, 20 °C)
Ignition Point 375 °C

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR