D-(-)-Quinic acid, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (1α,3R,4α,5R)-1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid

M-Clarity™ quality level = MQ200

8.00218.0050
8.00218.0050EA 968 DKK
8.00218.0050
D-(-)-Quinic acid, Sigma-Aldrich®
D-(-)-Quinic acid
Formel: C₇H₁₂O₆
Molekylvægt: 192.17 g/mol
MDL Number: MFCD00003864
CAS-nummer: 77-95-2
EINECS: 201-072-8

Bestil nu

Specifikationer Testresultater

Solubility 290 g/l (9 °C) (experimental)
Melting Point 162.5 °C
Molar Mass 192.17 g/mol
Bulk Density 670 - 750 kg/m3
Density 1.64 g/cm3 (20 °C)
pH 2.0 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR