Cell surface protein isolation and labelling kit, HOOK™
Leverandør: G Biosciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
786-316
786-316EA 5330 DKK
786-316
Cell surface protein isolation and labelling kit, HOOK™
Proteinekstraktions Protein Extraction Kits
The HOOK™ Cell Surface Protein Isolation Kit uses HOOK™ biotin labelling and purification technology in conjunction with Mammalian Cell PE LB™ Lysis Buffer to conveniently label cell surface proteins and isolate them for further analysis, including Western blotting. The Kit is supplied with all the necessary reagents and buffers for convenience and improved reproducibility. The kit is compatible with a wide variety of mammalian cells and can be used to compare treated and untreated cells and differences between different cell lines.
Bestillingsoplysninger: Accessories included. Additional supplies available on request, please contact your local Avantor sales office for further details.