Methylsalicylat, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Wintergreen Oil, Methyl 2-hydroxybenzoate

MQ100

76631-1ML-F 76631-5ML-F
76631-1ML-FEA 303 DKK
76631-1ML-F 76631-5ML-F
Methylsalicylat, Supelco®
Methylsalicylat
Formel: 2-(HO)C₆H₄CO₂CH₃
Molekylvægt: 152.15 g/mol
Kogepunkt: 432
Smeltepunkt: 100…101 °C
Massefylde: 1.184 g/cm³ (20 °C)
Flammepunkt: 99
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002214
CAS-nummer: 119-36-8
EINECS: 204-317-7
Merck Index: 13,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75c0f55c4fa148be5a24e67d720c70afd6188e55&vid=f0e08142a248049f2787cee081595ef3b709a252&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR