Electrostatic double pendulum
Leverandør: CORNELSEN EXPERIMENTA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
50080
765-0180EA 0 DKK
765-0180
Electrostatic double pendulum
Undervisningsmateriale Fysik undervisningsmateriale Electricity/Magnetism Learning Activities
Used to detect electric charge. Also suitable for use with belt generator and Faraday's cage. Equipped with connection for 4 mm banana plug.
  • Dimensions: total height 150 mm, cylinders: 10 × 10 mm