Resonance apparatus
Leverandør: CORNELSEN EXPERIMENTA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
46200
765-0156EA 0 DKK
765-0156
Resonance apparatus
Undervisningsmateriale Fysik undervisningsmateriale Waves and Oscillations Learning Activities
For acoustic detection of standing sound waves and for determining the wavelength of the sound source and then the speed of sound in air (Velocity=frequency×wavelength). The system is filled with water and a sound source (tuning fork) is held to the open tube end. By varying the height of the reservoir and the height of the liquid in the tube students can determine the wavelength using audible effects associated with standing waves.
  • Consisting of a glass tube with scale and three clamping clips connected via a hose and hose clamp to a liquid reservoir