Coffein, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine

M-Clarity™ quality level = MQ100

75035-5G-F
75035-5G-FEA 372 DKK
75035-5G-F
Coffein, Supelco®
Coffein
Formel: C₈H₁₀N₄O₂
Molekylvægt: 194.19 g/mol
Smeltepunkt: 235…239 °C
Massefylde: 1.23 g/cm³ (19 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005758
CAS-nummer: 58-08-2
EINECS: 200-362-1
Merck Index: 13,01636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e768db64f5fd9822647a6bd52ad29b93f1a315fa&vid=e875375a0b064eae42b139e3a9491d42d6180a3a&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR