Robot-spidser til Zymark® (Presto, RapidPlate, SciClone)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
923-262 921-262 921-261 921-021 923-261 923-021 920-262 920-261 920-021
737-2002EA 0 DKK
737-2002 737-2000 736-0102 736-0023 737-2001 736-0024 613-1865 613-1885 613-1886
Robot-spidser til Zymark® (Presto, RapidPlate, SciClone)
Pipettespidser
Højkvalitetsspidser, særligt velegnede til forskning i molekylærbiologiske laboratorier.
  • Transparent PP
  • Certified free of RNase, DNase, DNA and pyrogens
  • Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
  • Sterile tips: Sterilised by gamma radiation