Reagent containers for Microlab® STAR line
Leverandør: HAMILTON ROBOTICS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
194052 56694-01 56694-02 56695-01 56695-02 194051 182703
736-0834EA 657.13 DKK
736-0834 736-0831 736-0832 736-0835 736-0836 736-0830 736-0833
Reagent containers for Microlab® STAR line
Reagens reservoirs
PP, white or black. These reagent reservoirs with 60, 120 or 200 ml capacity can be placed in a reagent carrier on the STAR line instruments. The reservoirs are available self-standing or non-self-standing.
  • Special bottom geometry minimises residual volumes
  • With volume markings, without or with lid
  • With one (60 ml), three (120 ml) or four (200 ml) wave breakers to avoid splashes during movements of the filled containers
  • IVD compliant
  • Autoclavable
Bestillingsoplysninger: The carriers are not included and need to be ordered separately.