Robotic tips (CO-RE Tips) and consumables for Microlab
Leverandør: HAMILTON ROBOTICS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
235900 235937 235902 235904 235966 235985 184020 184022 235939 235965 235950 235679 235447 235444 235446 235449 235451 235971 235974 235978 235979 235983 235987 235695 235993 235989 235901 235903 235905 235936 235938 235940 235947 235948 235949 235935 235646 235647 235648 235452 235677 235678 235694 184021 184023 194050 194053 235806 235541 235688 235843 235844 235845 235846
736-0694EA 3381.8 DKK
736-0694 736-0788 736-0786 736-0793 736-0770 736-0792 736-0797 736-0799 736-0795 736-0791 736-0790 736-0811 736-0777 736-0804 736-0776 736-0805 736-0806 736-0771 736-0803 736-0772 736-0773 736-0774 736-0775 736-0779 736-0781 736-0780 736-0695 736-0787 736-0794 736-0765 736-0789 736-0796 736-0766 736-0767 736-0768 736-0764 736-0782 736-0783 736-0784 736-0807 736-0809 736-0810 736-0778 736-0798 736-0800 736-0801 736-0802 736-0785 736-0808 736-0853 HAMR235843 HAMR235844 HAMR235845 HAMR235846
Robotic tips (CO-RE Tips) and consumables for Microlab
Pipettespidser
Hamilton CO-RE tips were designed for MICROLAB pipetting workstations in order to solve the problem of sealing and alignment plagued by press-fit tip attachment techniques. Each CO-RE disposable tip has a circular groove inside the attachment orifice. An o-ring on the pipetting channel is expanded to fit this groove precisely providing an air-tight seal and straight alignment between tip and pipetting channel.
  • Pressure-based Liquid Level Detection (pLLD) and capacitive LLD (cLLD) with conductive (black) version
  • DNase-/RNase-free, pyrogen-free (endotoxin-