Vakuumfiltreringssystemer på flasketop, cellulosenitratmembran, Rapid-Flow™ Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
121-0020 121-0045 125-0020 125-0045 125-0080 126-0020 126-0045 126-0080 127-0020 127-0045 127-0080 450-0045 450-0080 450-0020
734-5001EA 1090 DKK
734-5001 734-5002 734-5003 734-5004 734-5005 734-5006 734-5007 734-5008 734-5009 734-5010 734-5011 734-5083 734-5084 734-5082
Vakuumfiltreringssystemer på flasketop, cellulosenitratmembran, Rapid-Flow™ Nalgene®
Filtre Topfiltre til flasker
Rapid-Flow™ disposable filter units with cellulose nitrate membrane, PS funnel and collection reservoir feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput.
  • Color coded according to membrane
  • Membrane with excellent wetting properties and fastest flow rates with aqueous solutions
  • Funnel and receiver flask graduated
  • Quick disconnect tubing adapter with cover
Sterile disposable filters for cold sterilisation, clarification, prefiltration and ultrafiltration of buffers, culture media and aqueous solutions.