Cell culture vessels, Corning® HYPERflask™ M, CellBIND® treated
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10024 10020 10030 10034 10031
734-4010EA 21390 DKK
734-4010 734-1491 734-1492 734-1493 CORN10031
Cell culture vessels, Corning® HYPERflask™ M, CellBIND® treated
Kolber Celledyrkningskolber
Optically clear polystyrene flasks feature a straight neck, flat-top cap, with or without bar code for use manually or with automated systems. No refrigeration or special handling required.
These exceptionally high-yield, high-performance flasks utilise a multilayered gas permeable growing surface for efficient gas exchange. Unique design allows flasks to produce 10× higher yields than standard flasks, increasing productivity and capacity as well as reducing processing time and incubator space. Ten interconnected polystyrene surfaces provide 1720 cm² area for cell growth with an air gapMore Product Information