96-Well Polypropylene Storage Blocks, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3956 3957 3960 3961 3959 3958
734-1675EA 3480 DKK
734-1675 734-1676 734-1679 734-1680 734-1678 734-1677
96-Well Polypropylene Storage Blocks, Corning®
Mikroplader HTS mikroplader
Plates are available in V-bottom and round-bottom formats to meet a wide variety of assay and storage needs.
Robolid combines the sealing ability of a storage mat with the rigidity of a plastic lid and is designed for repeated application and removal by automation and to prevent short-term evaporation. Robolid also features silicone sealing plugs for organic solvent resistance and low extractables.
All sterile products are certified RNase- and DNase-free. Built to the industry standard footprint.
Bestillingsoplysninger: Storage mats and sealing tapes sold separately.