Serological pipettes, Sterilin®
Leverandør: STERILIN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
47105 47110 40125 40105
734-0476EA 1520 DKK
734-0476 734-0477 612-3267 734-0463
Serological pipettes, Sterilin®
Pipetter Serologiske pipetter
PS, transparent, disposable, sterile.
  • Clear black printed graduations for maximum clarity, colour coded for easy identification, printed with product code/lot number and expiry date
  • Negative graduations for extra capacity
  • Gamma irradiated to ensure sterility and certified both non pyrogenic (<0,1 EU/ml) and non haemolytic
Bestillingsoplysninger: Batch specific certificate supplied in each case.
Advarsel: *Without suction adapter