Blue light transilluminator, TruBlu™
Leverandør: EDVOTEK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
557
733-2005EA 3580 DKK
733-2005
Blue light transilluminator, TruBlu™
Lysbord Blåt lys Transilluminatorer
The TruBlu™ blue light transilluminator is ideal for viewing DNA gels stained with SYBR® Safe, thus eliminating the need for UV light or ethidium bromide. It is optimised to fit Edvotek® gels as well as any other agarose gels.
  • Viewing area 270×150 mm
  • Optimised for all gels
  • Blue light high intensity control
  • Orange contrast lid
  • Most vivid results in education
The high intensity control and orange lid ensure superior visualisation.