AccuGENE® 1M Tris HCl buffer
Leverandør: LONZA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
51236 51237 51238
733-1652EA 1440 DKK
733-1652 733-1653 733-1654
AccuGENE® 1M Tris HCl buffer
Buffere Biologiske buffere
AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs
Tris-HCl is a solution containing tris base mixed with hydrocholoric acid. Tris has a pKa of 8,07 at STP and is used in bufferings solutions to prevent large changes in pH level. A staple of electrophoretic and other biological applications, Tris has a useful buffer range of 7 to 9, which coincides with the physiological pH typical of most living organisms. Tris is the 'T' in TAE and TBE Buffers.
Each reagent is prepared with 18 megOhm water, filtered using a 0,2 micron filter, and filled into sterile bottles.
Store all reagents at 18 to 26°C